Gene Transcripts

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
Chromosome Location
Chr. 19: 49335314 - 49340608 on Build GRCh38
UniGene ID
Hs.166556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040031.1 NP_001035120.1 6 659
NM_001774.2 NP_001765.1 6 693
XM_005259435.3 XP_005259492.1 6 695
XM_005259436.3 XP_005259493.1 6 691
XM_011527542.2 XP_011525844.1 6 656
XM_011527543.2 XP_011525845.1 6 761
XM_011527544.2 XP_011525846.1 6 730
XM_017027513.1 XP_016883002.1 6 691
GenBank mRNA AK058093.1 - 7 1040
AK301016.1 - 6 683
AK301328.1 - 6 686
AK316348.1 - 6 583
AL833693.1 - 5 1737
BC106752.1 - 6 610 40023348
BC110379.1 - 6 645 5752290
BT019851.1 - 6 572
CR541877.1 - 6 572
X14046.1 - 6 635

View Full Product Details