Gene Transcripts

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
Chromosome Location
Chr. 14: 99737699 - 99942060 on Build GRCh38
UniGene ID
Hs.12451
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008707.1 NP_001008707.1 6 662
NM_004434.2 NP_004425.2 5 605
XM_005267397.1 XP_005267454.1 5 675
XM_005267398.3 XP_005267455.1 5 608
XM_005267399.4 XP_005267456.2 5 647
XM_005267400.4 XP_005267457.1 5 631
XM_011536540.2 XP_011534842.1 5 678
XM_011536541.1 XP_011534843.1 5 688
XM_011536542.2 XP_011534844.1 5 647
XM_017021074.1 XP_016876563.1 5 675
GenBank mRNA AK126980.1 - 7 618
AK299812.1 - 6 941
AK307925.1 - 5 598
BC032541.1 - 5 581 5533599
BC033043.1 - 5 605 4821202
BX247979.1 - 6 674
BX420178.2 - 6 674
U97018.1 - 5 604

View Full Product Details