Gene Transcripts

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
Chromosome Location
Chr. 9: 120952335 - 121075174 on Build GRCh38
UniGene ID
Hs.494997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317163.1 NP_001304092.1 12 1746
NM_001317164.1 NP_001304093.1 12 1417
NM_001735.2 NP_001726.2 12 1420
XM_011518980.2 XP_011517282.1 13 1789
XM_017015102.1 XP_016870591.1 14 1941
XM_017015103.1 XP_016870592.1 12 1574
XM_017015104.1 XP_016870593.1 14 1941
GenBank mRNA AB209031.1 - 12 1746
AK310774.1 - 12 1417
AK310780.1 - 12 1419
BC022299.1 - 12 1421
BC113738.1 - 12 1415 8322790
BC113740.1 - 12 1415 8322792
M57729.1 - 12 1402
M65134.1 - 2 157

View Full Product Details