Gene Transcripts

Gene Symbol
KLHDC10
Entrez Gene ID
23008
Gene Name
kelch domain containing 10
Gene Aliases
slim
Chromosome Location
Chr. 7: 130070509 - 130135720 on Build GRCh38
UniGene ID
Hs.520710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014997.3 NP_055812.1 3 529
XM_005250233.4 XP_005250290.1 2 418
GenBank mRNA BC036464.2 - 3 511
BC044884.1 - 3 531 5273087
D87454.1 - 3 271

View Full Product Details