Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079670.2 | NP_001073138.1 | 8 | 969 | |
NM_001287337.1 | NP_001274266.1 | 9 | 1107 | ||
NM_001287338.1 | NP_001274267.1 | 9 | 1102 | ||
NM_001287339.1 | NP_001274268.1 | 7 | 771 | ||
NM_030925.3 | NP_112187.2 | 6 | 1020 | ||
XM_011535254.2 | XP_011533556.1 | 10 | 1511 | ||
XM_011535255.2 | XP_011533557.1 | 8 | 991 | ||
XM_011535256.2 | XP_011533558.1 | 9 | 1256 | ||
XM_017020785.1 | XP_016876274.1 | 9 | 1383 | ||
XM_017020786.1 | XP_016876275.1 | 10 | 1520 | ||
XM_017020787.1 | XP_016876276.1 | 10 | 1773 | ||
XM_017020788.1 | XP_016876277.1 | 11 | 1649 | ||
GenBank mRNA | AA278473.1 | - | 4 | 456 | |
AK022639.1 | - | 5 | 565 | ||
AK309011.1 | - | 3 | 295 | ||
AY288977.2 | - | 9 | 1034 | ||
BC010993.2 | - | 6 | 845 | 3956127 | |
BX647518.1 | - | 8 | 906 |