Gene Transcripts

Gene Symbol
OR5P3
Entrez Gene ID
120066
Gene Name
olfactory receptor family 5 subfamily P member 3
Gene Aliases
JCG1
Chromosome Location
Chr. 11: 7825037 - 7825972 on Build GRCh38
UniGene ID
Hs.351825
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153445.1 NP_703146.1 1 121
GenBank mRNA AF158377.1 - 1 121

View Full Product Details