Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130046.1 | NP_001123518.1 | 4 | 435 | |
NM_004591.2 | NP_004582.1 | 4 | 438 | ||
GenBank mRNA | BC020698.1 | - | 4 | 435 | |
D86955.1 | - | 4 | 426 | ||
U64197.1 | - | 4 | 407 | ||
U77035.1 | - | 4 | 368 |