Gene Transcripts
- Gene Symbol
- Pnn
- Entrez Gene ID
- 18949
- Gene Name
- pinin
- Gene Aliases
- AU045199, D12Ertd512e
- Chromosome Location
- Chr. 12: 59066919 - 59074017 on Build GRCm38
- UniGene ID
- Mm.22347
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008891.2 |
NP_032917.2
|
2, 3 |
261 |
|
GenBank mRNA |
AK009055.1 |
- |
2, 3 |
216 |
|
AK028219.1 |
- |
2, 3 |
202 |
|
AK030978.1 |
- |
2, 3 |
190 |
|
AK050669.1 |
- |
2, 3 |
207 |
|
AK051640.1 |
- |
2, 3 |
259 |
|
AK157587.1 |
- |
2, 3 |
205 |
|
AK160616.1 |
- |
2, 3 |
196 |
|
AK161109.1 |
- |
2, 3 |
201 |
|
BC132571.1 |
- |
2, 3 |
195 |
40130848 |
BC138014.1 |
- |
2, 3 |
195 |
8861033 |
BY775218.1 |
- |
2, 3 |
262 |
|
Y08701.1 |
- |
2, 3 |
195 |
|
View Full Product Details