Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004666.2 | NP_004657.2 | 1 | 212 | |
GenBank mRNA | AJ132099.1 | - | 1 | 206 | |
AK290425.1 | - | 1 | 209 | ||
AK314265.1 | - | 1 | 209 | ||
BC096265.3 | - | 1 | 206 | 40006204 | |
BC096266.3 | - | 1 | 206 | 40006205 | |
BC096267.3 | - | 1 | 206 | 40006207 | |
BC096268.1 | - | 1 | 205 | 40006208 | |
BJ995136.1 | - | 1 | 212 | ||
U39664.1 | - | 1 | 207 |