Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 15 | 3157 | |
GenBank mRNA | AK028528.1 | - | 1 | 1035 | |
AK032117.1 | - | 1 | 738 | ||
AK033912.1 | - | 1 | 760 | ||
AK034443.1 | - | 1 | 283 | ||
AK035962.1 | - | 6 | 1568 | ||
AK044922.1 | - | 1 | 51 | ||
AK082492.1 | - | 1 | 766 | ||
AK136861.1 | - | 15 | 3125 | ||
AK138155.1 | - | 1 | 670 | ||
AK149055.1 | - | 4 | 1297 | ||
AK196191.1 | - | 1 | 74 | ||
AK201776.1 | - | 1 | 74 | ||
BC020178.1 | - | 15 | 3054 | 4507346 | |
BC062941.1 | - | 15 | 3077 |