Gene Transcripts

Gene Symbol
Pikfyve
Entrez Gene ID
18711
Gene Name
phosphoinositide kinase, FYVE finger containing
Gene Aliases
5230400C17Rik, Pip5k, Pip5k3, Pipk5k3, PipkIII, p235
Chromosome Location
Chr. 1: 65186639 - 65278696 on Build GRCm38
UniGene ID
Mm.38370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011086.2 NP_035216.2 7 1000
GenBank mRNA AF102777.1 - 7 980
AK080527.1 - 6 901
AK139116.1 - 6 915
AK143565.1 - 6 947
AK165350.1 - 6 961
AK173077.1 - 6 902

View Full Product Details