Gene Transcripts

Gene Symbol
Prl6a1
Entrez Gene ID
19111
Gene Name
prolactin family 6, subfamily a, member 1
Gene Aliases
PLP-B, Prlpb
Chromosome Location
Chr. 13: 27312627 - 27319252 on Build GRCm38
UniGene ID
Mm.7512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011166.2 NP_035296.1 2 196
GenBank mRNA AF011384.1 - 2 195
AF015563.1 - 2 196

View Full Product Details