Gene Transcripts

Gene Symbol
Sod3
Entrez Gene ID
20657
Gene Name
superoxide dismutase 3, extracellular
Gene Aliases
AI314465, EC-SOD
Chromosome Location
Chr. 5: 52363804 - 52369738 on Build GRCm38
UniGene ID
Mm.2407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011435.3 NP_035565.1 2 1758
GenBank mRNA AK075570.1 - 2 1613
AK145194.1 - 1617
BC010975.1 - 2 1576 3983917
U38261.1 - 2 1759

View Full Product Details