Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011481.3 | NP_035611.3 | 7 | 1640 | |
GenBank mRNA | AK049582.1 | - | 7 | 901 | |
BC120633.1 | - | 7 | 1619 | 40129556 | |
BC125325.1 | - | 7 | 1616 | 40129840 | |
D26186.1 | - | 7 | 1486 | ||
D49427.1 | - | 7 | 1654 |