Gene Transcripts

Gene Symbol
Lgi4
Entrez Gene ID
243914
Gene Name
leucine-rich repeat LGI family, member 4
Gene Aliases
Lgil3, clp
Chromosome Location
Chr. 7: 31058953 - 31070935 on Build GRCm38
UniGene ID
Mm.389870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144556.2 NP_653139.2 9 3046
GenBank mRNA AJ487521.1 - 9 3047
AJ487960.1 - 9 3046
AK033967.1 - 9 2864
AK134643.1 - 9 3640
BC098469.1 - 4 2552 30099294

View Full Product Details