Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146211.3 | NP_666323.2 | 12 | 2189 | |
GenBank mRNA | AK035788.1 | - | 12 | 2067 | |
AK036165.1 | - | 12 | 2192 | ||
AK076103.1 | - | 12 | 2046 | ||
AK154775.1 | - | 12 | 2043 | ||
AK188359.1 | - | 1 | 246 | ||
BC027801.1 | - | 4 | 1882 | ||
BC032165.1 | - | 12 | 2221 | 5353334 | |
BC032185.1 | - | 3 | 1003 | ||
BC034208.1 | - | 9 | 1507 | ||
BC056951.1 | - | 12 | 2080 | 5695490 |