Gene Transcripts

Gene Symbol
Fam63b
Entrez Gene ID
235461
Gene Name
family with sequence similarity 63, member B
Gene Aliases
AI465301, B230380D07Rik, mKIAA1164
Chromosome Location
Chr. 9: 70599014 - 70657296 on Build GRCm38
UniGene ID
Mm.170855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172772.2 NP_766360.2 9 2391
GenBank mRNA AK041072.1 - 1 29
AK046403.1 - 9 2378
AK077470.1 - 6 1323
BC049357.1 - 9 2364
BC055816.1 - 9 2119
BC058683.1 - 9 2349 6834871

View Full Product Details