Gene Transcripts

Gene Symbol
Zcchc7
Entrez Gene ID
319885
Gene Name
zinc finger, CCHC domain containing 7
Gene Aliases
4930572I07Rik, D4Wsu132e
Chromosome Location
Chr. 4: 44756559 - 44932215 on Build GRCm38
UniGene ID
Mm.331576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138590.4 NP_613056.2 2 406
GenBank mRNA AK029808.1 - 2 413
AK032353.1 - 2 378
AK039158.1 - 2 327
AK043238.1 - 2 319
AK080785.1 - 2 319
AK157728.1 - 2 309
BC050200.1 - 1 281
BC117020.1 - 2 309 40126339
BC117022.1 - 2 309 40126341
BC144774.1 - 2 313 9053322
CJ155194.1 - 2 378

View Full Product Details