Gene Transcripts

Gene Symbol
Hist3h2a
Entrez Gene ID
319162
Gene Name
histone cluster 3, H2a
Gene Aliases
Chromosome Location
Chr. 11: 58954685 - 58955192 on Build GRCm38
UniGene ID
Mm.212549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK051448.1 - 925
AK082083.1 - 928
AK087537.1 - 926
BC058119.1 - 897 6819179
BC063781.1 - 891 6493838

View Full Product Details