Gene Transcripts

Gene Symbol
Ltb4r1
Entrez Gene ID
16995
Gene Name
leukotriene B4 receptor 1
Gene Aliases
BLT1, BLTR, Ltb4r, mBLTR
Chromosome Location
Chr. 14: 55765050 - 55768494 on Build GRCm38
UniGene ID
Mm.20853
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008519.2 NP_032545.1 2 1373
GenBank mRNA AF044030.1 - 2 1369
AF110104.1 - 2 1320
AK010895.1 - 2 1359
AK155181.1 - 3 1166
BC064063.1 - 2 1304 874601

View Full Product Details