Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010722.5 | NP_034852.2 | 6 | 986 | |
GenBank mRNA | AK132671.1 | - | 3 | 694 | |
AK199324.1 | - | 1 | 38 | ||
BC042430.1 | - | 6 | 991 | 3674002 | |
BC051985.1 | - | 6 | 962 | 5695459 | |
D13455.1 | - | 3 | 673 | ||
X54098.1 | - | 6 | 945 |