Gene Transcripts

Gene Symbol
Lrrn3
Entrez Gene ID
16981
Gene Name
leucine rich repeat protein 3, neuronal
Gene Aliases
NLRR-3
Chromosome Location
Chr. 12: 41451668 - 41486057 on Build GRCm38
UniGene ID
Mm.404786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271708.1 NP_001258637.1 3 1269
NM_001271709.1 NP_001258638.1 2 1235
NM_010733.3 NP_034863.1 2 1240
GenBank mRNA AK036316.1 - 2 1138
BC069041.1 - 2 934 5698788
D49802.1 - 2 1151

View Full Product Details