Gene Transcripts
- Gene Symbol
- PCBP2
- Entrez Gene ID
- 5094
- Gene Name
- poly(rC) binding protein 2
- Gene Aliases
- HNRNPE2, HNRPE2, hnRNP-E2
- Chromosome Location
- Chr. 12: 53452102 - 53481162 on Build GRCh38
- UniGene ID
- Hs.546271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098620.2 |
NP_001092090.1
|
6, 7 |
721 |
|
NM_001128911.1 |
NP_001122383.1
|
6, 7 |
721 |
|
NM_001128912.1 |
NP_001122384.1
|
6, 7 |
721 |
|
NM_001128913.1 |
NP_001122385.1
|
6, 7 |
721 |
|
NM_001128914.1 |
NP_001122386.1
|
6, 7 |
721 |
|
NM_005016.5 |
NP_005007.2
|
6, 7 |
721 |
|
NM_031989.4 |
NP_114366.1
|
6, 7 |
721 |
|
GenBank mRNA |
AB188306.1 |
- |
6, 7 |
721 |
|
AB208825.1 |
- |
5, 6 |
455 |
|
AK225357.1 |
- |
5, 6 |
435 |
|
AK292141.1 |
- |
6, 7 |
498 |
|
AK299210.1 |
- |
4, 5 |
315 |
|
AK302067.1 |
- |
5, 6 |
438 |
|
BC001155.1 |
- |
5, 6 |
459 |
3356341 |
BC035420.1 |
- |
6, 7 |
694 |
|
BC071942.1 |
- |
6, 7 |
711 |
6424433 |
BC107688.1 |
- |
5, 6 |
387 |
5395926 |
CX164318.1 |
- |
6, 7 |
713 |
|
X78136.1 |
- |
5, 6 |
393 |
|
View Full Product Details