Gene Transcripts

Gene Symbol
H2-T3
Entrez Gene ID
15043
Gene Name
histocompatibility 2, T region locus 3
Gene Aliases
H-2T3, H2-Tw3, TL
Chromosome Location
Chr. 17: 36185571 - 36190151 on Build GRCm38
UniGene ID
Mm.426002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008208.4 NP_032234.3 6 1987
GenBank mRNA AK033602.1 - 6 1988

View Full Product Details