Gene Transcripts

Gene Symbol
Gab2
Entrez Gene ID
14389
Gene Name
growth factor receptor bound protein 2-associated protein 2
Gene Aliases
AI463667, D130058I17Rik, p97
Chromosome Location
Chr. 7: 97081751 - 97308951 on Build GRCm38
UniGene ID
Mm.42033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162477.1 NP_001155949.1 2 358
NM_010248.2 NP_034378.2 2 358
GenBank mRNA AB018414.1 - 2 283
AF104244.1 - 2 358
BC103524.1 - 2 333 40039291
BC103525.1 - 2 333 40039280
BC103526.1 - 2 333 40039281
BC103527.1 - 2 333 40039282

View Full Product Details