Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007691.5 | NP_031717.2 | 11 | 1256 | |
GenBank mRNA | AF016583.1 | - | 10 | 1165 | |
AK011258.1 | - | 11 | 1200 | ||
AK033179.1 | - | 11 | 1226 | ||
AK045336.1 | - | 11 | 1198 | ||
AK166675.1 | - | 11 | 1257 | ||
BC037613.1 | - | 10 | 969 | ||
BC053394.1 | - | 11 | 1193 | ||
BC064445.1 | - | 10 | 1655 | ||
BC100386.1 | - | 10 | 1331 | 30481390 |