Gene Transcripts

Gene Symbol
Clk4
Entrez Gene ID
12750
Gene Name
CDC like kinase 4
Gene Aliases
AI987988, C85119
Chromosome Location
Chr. 11: 51263167 - 51281770 on Build GRCm38
UniGene ID
Mm.239354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF033566.1 - 2 219
AY653308.1 - 1 141
BC012675.1 - 2 241 3982920
CT010293.1 - 1 141

View Full Product Details