Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007725.2 | NP_031751.1 | 3, 4 | 335 | |
GenBank mRNA | AK051845.1 | - | 3, 4 | 336 | |
AK189304.1 | - | 3, 4 | 263 | ||
AK218047.1 | - | 1, 2 | 29 | ||
AK218463.1 | - | 2, 3 | 197 | ||
BC008181.1 | - | 3, 4 | 253 | 3589470 | |
BC009144.1 | - | 3, 4 | 290 | 3594856 | |
BC018482.1 | - | 3, 4 | 305 | 3589158 |