Gene Transcripts
- Gene Symbol
- Apbb1
- Entrez Gene ID
- 11785
- Gene Name
- amyloid beta (A4) precursor protein-binding, family B, member 1
- Gene Aliases
- Fe65, Rir
- Chromosome Location
- Chr. 7: 105558465 - 105581653 on Build GRCm38
- UniGene ID
- Mm.38469
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253885.1 |
NP_001240814.1
|
6, 7 |
1475 |
|
NM_001253886.1 |
NP_001240815.1
|
6, 7 |
1475 |
|
NM_001253887.1 |
NP_001240816.1
|
6, 7 |
475 |
|
NM_009685.3 |
NP_033815.1
|
6, 7 |
1289 |
|
GenBank mRNA |
AF206720.1 |
- |
6, 7 |
1198 |
|
AK030748.1 |
- |
6, 7 |
1172 |
|
AK075730.1 |
- |
6, 7 |
455 |
|
AK083830.1 |
- |
6, 7 |
1205 |
|
AK144340.1 |
- |
6, 7 |
1476 |
|
AK164140.1 |
- |
6, 7 |
1200 |
|
BC036956.1 |
- |
5, 6 |
1113 |
|
BC048395.1 |
- |
4, 5 |
621 |
6392016 |
View Full Product Details