Gene Transcripts

Gene Symbol
Apbb1
Entrez Gene ID
11785
Gene Name
amyloid beta (A4) precursor protein-binding, family B, member 1
Gene Aliases
Fe65, Rir
Chromosome Location
Chr. 7: 105558465 - 105581653 on Build GRCm38
UniGene ID
Mm.38469
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253885.1 NP_001240814.1 2 526
NM_001253886.1 NP_001240815.1 2 526
NM_009685.3 NP_033815.1 2 340
GenBank mRNA AF206720.1 - 2 249
AK030748.1 - 2 223
AK083830.1 - 2 256
AK144340.1 - 2 527
AK164140.1 - 2 251
BC036956.1 - 1 164
CX224409.1 - 2 340

View Full Product Details