Gene Transcripts

Gene Symbol
Apbb2
Entrez Gene ID
11787
Gene Name
amyloid beta (A4) precursor protein-binding, family B, member 2
Gene Aliases
2310007D03Rik, FE65L1, Rirl1, TR2L, Zfra
Chromosome Location
Chr. 5: 66298725 - 66618817 on Build GRCm38
UniGene ID
Mm.5159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201413.1 NP_001188342.1 5 1058
NM_001201414.1 NP_001188343.1 5 1121
NM_001201415.1 NP_001188344.1 5 1058
NM_009686.2 NP_033816.1 5 1058
GenBank mRNA AK004792.1 - 5 912
AK032770.1 - 5 924
AK153260.1 - 5 1087
AK167649.1 - 5 923
AK169495.1 - 5 911
AK169959.1 - 5 999
BC076587.1 - 5 904 30550087

View Full Product Details