Gene Transcripts

Gene Symbol
HMGB3
Entrez Gene ID
3149
Gene Name
high mobility group box 3
Gene Aliases
HMG-2a, HMG-4, HMG2A, HMG4
Chromosome Location
Chr. X: 150980508 - 150990775 on Build GRCh38
UniGene ID
Hs.19114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301228.1 NP_001288157.1 3 415
NM_001301229.1 NP_001288158.1 3 306
NM_001301231.1 NP_001288160.1 3 345
NM_005342.3 NP_005333.2 3 310
GenBank mRNA BC037210.1 - 2 215
BC070482.1 - 4 394 6140626
BF979310.1 - 3 351
BX537505.1 - 3 295
DB191695.1 - 3 295
DN996967.1 - 3 317
L05085.1 - 2 215
Y10043.1 - 3 289

View Full Product Details