Gene Transcripts

Gene Symbol
Dok1
Entrez Gene ID
13448
Gene Name
docking protein 1
Gene Aliases
AW557123, p62DOK
Chromosome Location
Chr. 6: 83030932 - 83033471 on Build GRCm38
UniGene ID
Mm.156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291799.1 NP_001278728.1 5 1796
NM_010070.4 NP_034200.4 5 1798
GenBank mRNA BC013066.1 - 5 1791
BY645588.1 - 1 394

View Full Product Details