Gene Transcripts

Gene Symbol
Clcn4-2
Entrez Gene ID
12727
Gene Name
chloride channel 4-2
Gene Aliases
Clc4-2, Clcn4
Chromosome Location
Chr. 7: 7282309 - 7300851 on Build GRCm38
UniGene ID
Mm.297883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK035804.1 - 12 2552
AK050833.1 - 2 324
BC110668.1 - 13 2912 6391053

View Full Product Details