Gene Transcripts
- Gene Symbol
- Clns1a
- Entrez Gene ID
- 12729
- Gene Name
- chloride channel, nucleotide-sensitive, 1A
- Gene Aliases
- 2610036D06Rik, 2610100O04Rik, Clci, Clcni, ICLN
- Chromosome Location
- Chr. 7: 97696657 - 97720793 on Build GRCm38
- UniGene ID
- Mm.21482
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023671.2 |
NP_076160.1
|
6, 7 |
818 |
|
GenBank mRNA |
AK011691.1 |
- |
6, 7 |
798 |
|
AK011789.1 |
- |
6, 7 |
803 |
|
AK031877.1 |
- |
6, 7 |
800 |
|
AK078050.1 |
- |
6, 7 |
771 |
|
AK133981.1 |
- |
6, 7 |
811 |
|
AK151356.1 |
- |
6, 7 |
803 |
|
AK151944.1 |
- |
6, 7 |
803 |
|
AK169259.1 |
- |
6, 7 |
782 |
|
BC005575.1 |
- |
6, 7 |
784 |
3967933 |
View Full Product Details