Gene Transcripts

Gene Symbol
Sult1d1
Entrez Gene ID
53315
Gene Name
sulfotransferase family 1D, member 1
Gene Aliases
5033411P13Rik, AI987815, ST1d1, SULT-N, Sultn
Chromosome Location
Chr. 5: 87554650 - 87569006 on Build GRCm38
UniGene ID
Mm.6824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016771.3 NP_058051.3 8 2058
GenBank mRNA AK017162.1 - 2 1250
BC066190.1 - 8 2012 30472198

View Full Product Details