Gene Transcripts

Gene Symbol
Ddit4l
Entrez Gene ID
73284
Gene Name
DNA-damage-inducible transcript 4-like
Gene Aliases
1700037B15Rik, 1700108M02Rik, REDD2, RTP801L, Smhs1
Chromosome Location
Chr. 3: 137623672 - 137628332 on Build GRCm38
UniGene ID
Mm.250841
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030143.4 NP_084419.2 3 1509
GenBank mRNA AK029362.1 - 1 1076
AK053333.1 - 3 1446
AK075693.1 - 4 1400

View Full Product Details