Gene Transcripts

Gene Symbol
Dtx4
Entrez Gene ID
207521
Gene Name
deltex 4 homolog (Drosophila)
Gene Aliases
AI449438, BC044798, D930042H13, RNF155
Chromosome Location
Chr. 19: 12466329 - 12502201 on Build GRCm38
UniGene ID
Mm.247695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172442.3 NP_766030.3 2 1250
GenBank mRNA BC058647.1 - 2 708

View Full Product Details