Gene Transcripts

Gene Symbol
Hirip3
Entrez Gene ID
233876
Gene Name
HIRA interacting protein 3
Gene Aliases
B130036O03, C86302
Chromosome Location
Chr. 7: 126861972 - 126865377 on Build GRCm38
UniGene ID
Mm.363427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172746.3 NP_766334.1 7 2315
GenBank mRNA AK045122.1 - 7 1850
AK135526.1 - 7 1860
AK161657.1 - 7 1850
AK162096.1 - 4 877
BC055687.1 - 7 2315 6414655

View Full Product Details