Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102409.1 | NP_001095879.1 | 1 | 329 | |
NM_001102410.1 | NP_001095880.1 | 1 | 329 | ||
NM_201375.2 | NP_958763.1 | 1 | 329 | ||
GenBank mRNA | AY462056.1 | - | 1 | 245 | |
AY462057.1 | - | 1 | 245 | ||
AY462059.1 | - | 1 | 245 | ||
AY660572.1 | - | 1 | 245 | ||
BC108335.1 | - | 1 | 296 | 5124483 | |
BC108934.1 | - | 1 | 237 | ||
BC139720.1 | - | 1 | 237 | 40039854 | |
BY773346.1 | - | 1 | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128967.1 | - | 160 | Chr. - - [-] |