Gene Transcripts

Gene Symbol
Usp17lb
Entrez Gene ID
381944
Gene Name
ubiquitin specific peptidase 17-like B
Gene Aliases
DUB-1A, Dub1a
Chromosome Location
Chr. 7: 104840257 - 104842597 on Build GRCm38
UniGene ID
Mm.334094
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201409.2 NP_958811.2 2 714
GenBank mRNA AY257250.1 - 2 711
BC137942.1 - 2 810 8860961
BC145058.1 - 2 810 9053606

View Full Product Details