Gene Transcripts

Gene Symbol
Rhog
Entrez Gene ID
56212
Gene Name
ras homolog gene family, member G
Gene Aliases
2810426G09Rik, Arhg, Sid10750
Chromosome Location
Chr. 7: 102239123 - 102250118 on Build GRCm38
UniGene ID
Mm.259795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019566.3 NP_062512.1 2 684
GenBank mRNA AB025943.1 - 1 542
AK143623.1 - 2 677
AK149848.1 - 2 677
AK169853.1 - 2 668
AK188210.1 - 1 161
BC059775.1 - 2 662 5370916

View Full Product Details