Gene Transcripts

Gene Symbol
Unk
Entrez Gene ID
217331
Gene Name
unkempt homolog (Drosophila)
Gene Aliases
B230379M23Rik, Zc3h5, Zc3hdc5, mKIAA1753
Chromosome Location
Chr. 11: 116030295 - 116061214 on Build GRCm38
UniGene ID
Mm.297706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286006.1 NP_001272935.1 5, 6 827
NM_172569.4 NP_766157.1 5, 6 827
GenBank mRNA AK046395.1 - 5, 6 822
AK077635.1 - 5, 6 830
AK078095.1 - 5, 6 818
AK129438.1 - 5, 6 805
AK134361.1 - 5, 6 947
BC003195.1 - 2, 3 273
BC039964.1 - 5, 6 789
BC054452.1 - 5, 6 764 6491704

View Full Product Details