Gene Transcripts

Gene Symbol
Gadd45gip1
Entrez Gene ID
102060
Gene Name
growth arrest and DNA-damage-inducible, gamma interacting protein 1
Gene Aliases
2310040G17Rik, AI425883, Crif1, MRP-L59, Plinp1
Chromosome Location
Chr. 8: 84832282 - 84835482 on Build GRCm38
UniGene ID
Mm.489649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183358.4 NP_899202.3 2 614
GenBank mRNA AK002738.1 - 2 620
AK003314.1 - 2 615
AK009723.1 - 2 613
AK019570.1 - 2 615
AK028227.1 - 2 615
BC022592.1 - 2 499
BC061069.1 - 2 614 6740575

View Full Product Details