Gene Transcripts

Gene Symbol
NELFA
Entrez Gene ID
7469
Gene Name
negative elongation factor complex member A
Gene Aliases
NELF-A, P/OKcl.15, WHSC2
Chromosome Location
Chr. 4: 1982714 - 2009972 on Build GRCh38
UniGene ID
Hs.21771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005663.4 NP_005654.3 2 638
XM_017008589.1 XP_016864078.1 3 1459
GenBank mRNA AB044549.1 - 1 425
AF101434.1 - 2 367
AK001304.1 - 2 425
AK095737.1 - 1 681
AK126056.1 - 2 1513
AK222853.1 - 2 375
BC002764.2 - 2 378 3630594

View Full Product Details