Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253719.1 | NP_001240648.1 | 4 | 544 | |
NM_178673.4 | NP_848788.2 | 4 | 804 | ||
GenBank mRNA | AF374460.1 | - | 4 | 778 | |
AK033652.1 | - | 4 | 795 | ||
AK046070.1 | - | 4 | 545 | ||
AK081152.1 | - | 4 | 795 | ||
AK138388.1 | - | 4 | 1096 | ||
AK140611.1 | - | 4 | 807 | ||
BC075640.1 | - | 4 | 767 | 30545529 |