Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207214.3 | NP_997097.1 | 5 | 1036 | |
GenBank mRNA | AK134562.1 | - | 5 | 548 | |
AK152307.1 | - | 5 | 765 | ||
AK157473.1 | - | 5 | 731 | ||
AK164067.1 | - | 5 | 758 | ||
AK167145.1 | - | 5 | 724 | ||
AK170148.1 | - | 5 | 729 | ||
AK171012.1 | - | 5 | 754 | ||
BC049967.1 | - | 5 | 693 | 3672313 |