Gene Transcripts

Gene Symbol
Scgb1b2
Entrez Gene ID
57426
Gene Name
secretoglobin, family 1B, member 2
Gene Aliases
Abpa2, Abph, Apbh, LGP, Mja1l, Scgb1b1
Chromosome Location
Chr. 7: 31290519 - 31291816 on Build GRCm38
UniGene ID
Mm.6205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020563.3 NP_065588.1 3 332
GenBank mRNA AF008595.1 - 3 333
BC115490.1 - 3 333 40100845
BC115491.1 - 3 333 40100846

View Full Product Details