Gene Transcripts

Gene Symbol
ARPC3
Entrez Gene ID
10094
Gene Name
actin related protein 2/3 complex subunit 3
Gene Aliases
ARC21, p21-Arc
Chromosome Location
Chr. 12: 110434890 - 110450411 on Build GRCh38
UniGene ID
Hs.524741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278556.1 NP_001265485.1 5, 6 531
GenBank mRNA AF004561.1 - 4, 5 380
AF006086.1 - 5, 6 405
AK297309.1 - 5, 6 462
AK311789.1 - 5, 6 478
BC067747.1 - 5, 6 437 5302338
BC078162.1 - 5, 6 414 6148231
BG723870.1 - 5, 6 495
CR407667.1 - 4, 5 380
DQ328220.1 - 5, 6 461

View Full Product Details