Gene Transcripts

Gene Symbol
Pdzd7
Entrez Gene ID
100503041
Gene Name
PDZ domain containing 7
Gene Aliases
9130207N01, EG435601, Pdzk7
Chromosome Location
Chr. 19: 45026907 - 45058308 on Build GRCm38
UniGene ID
Mm.329877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195265.1 NP_001182194.1 16 3632
GenBank mRNA AK033653.1 - 6 1527
BC096488.1 - 6 1748

View Full Product Details